1-Fluor-2-ethinylbenzol 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
1-Ethinyl-2-fluorbenzol, (2-Fluorphenyl)ethin, (2-Fluorphenyl)acetylen
380990010
380992500
ACRO380990010EA
53
EUR
ACRO380990010
ACRO380992500
1-Fluor-2-ethinylbenzol 97%
1-Fluor-2-ethinylbenzol
Formel:
C₈H₅F Flammpunkt: 37 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00799540 CAS-Nummer: 766-49-4 UN: 1993 |
Spezifikation Testergebnisse
Appearance | Clear yellow to brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5240 to 1.5270 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...