1-Fluor-2-ethinylbenzol
Lieferant: FLUOROCHEM
Synonyme:
1-Ethinyl-2-fluorbenzol, (2-Fluorphenyl)ethin, (2-Fluorphenyl)acetylen
008806-1G
008806-5G
FLUO008806-1GEA
48.8
EUR
FLUO008806-1G
FLUO008806-5G
1-Fluor-2-ethinylbenzol
1-Fluor-2-ethinylbenzol
Formel:
C₈H₅F Lagertemperatur: Kühlschrank |
MDL:
MFCD00799540 CAS-Nummer: 766-49-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...