1-Fluor-4-ethinylbenzol ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (4-Fluorphenyl)acetylen, 1-Ethinyl-4-fluorbenzol, (4-Fluorphenyl)ethin

H26269.03 H26269.14 H26269.06
H26269.03EA 55.6 EUR
H26269.03 H26269.14 H26269.06
1-Fluor-4-ethinylbenzol ≥99%
1-Fluor-4-ethinylbenzol

Purity: 99% CAS No.: 766-98-3 Molecular Formula: C8H5F Formula Weight: 120.12 Flash Point: 29°(85°F) Refractive Index: 1.5170 Odour: Characteristic Melting Point: 27-28° Boiling Point: 55-56°/40mm Beilstein Reference: 1099285 UN No.: 1325 MDL No.: MFCD00168823

Formel: C₈H₅F
Molecular Weight: 120.13 g/mol
Siedepunkt: 55…56 °C (40 mmHg)
Schmelzpunkt: 27…28 °C
Dichte: 1.048
Flammpunkt: 29 °C (85 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00168823
CAS-Nummer: 766-98-3
UN: 1325
ADR: 4.1 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR