4-Ethinylanisol ≥99%
Lieferant: Thermo Scientific
Synonyme:
1-Ethinyl-4-methoxybenzol, (4-Methoxyphenyl)acetylen, 1-Methoxy-4-ethinylbenzol, (4-Methoxyphenyl)ethin
H26283.03
H26283.06
H26283.03EA
62.8
EUR
H26283.03
H26283.06
4-Ethinylanisol ≥99%
4-Ethinylanisol
Purity: 99%
CAS No.: 768-60-5
Molecular Formula: C9H8O
Formula Weight: 132.16
Flash Point: 82°(180°F)
Refractive Index: 1.563
Melting Point: 27-29°
Boiling Point: 87-91°/11mm
MDL No.: MFCD00168815
Formel:
C₉H₈O Molecular Weight: 132,16 g/mol Siedepunkt: 87…91 °C (11 mmHg) Schmelzpunkt: 27…29 °C Dichte: 1,019 Flammpunkt: 82 °C (180 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00168815 CAS-Nummer: 768-60-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...