Phenylacetylen ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Phenylethin, Ethinylbenzol
A12139.14
A12139.22
A12139.36
A12139.14EA
34.9
EUR
A12139.14
A12139.22
A12139.36
Phenylacetylen ≥98%
Phenylacetylen
Purity: 98+%
CAS No.: 536-74-3
Molecular Formula: C8H6
Formula Weight: 102.14
Flash Point: 27°(81°F)
Refractive Index: 1.5480
Melting Point: -45°
Boiling Point: 142-144°
Beilstein Reference: 605461
Merck Reference: 14,3861
UN No.: 3295
MDL No.: MFCD00008570
Formel:
C₈H₆ Molecular Weight: 102,14 g/mol Siedepunkt: 142…144 °C Schmelzpunkt: –45 °C Dichte: 0,930 Flammpunkt: 27 °C (81 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008570 CAS-Nummer: 536-74-3 EINECS: 208-645-1 UN: 3295 ADR: 3,III Merck Index: 14,3861 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...