4-Ethylphenylboronsäure 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: (4-Ethylphenyl)metaborsäure, 4-Ethylbenzolboronsäure, Dihydroxy(4-ethylphenyl)boran

AC35932.1 AC35932.10
MAYBAC35932.1EA 0 EUR
MAYBAC35932.1 MAYBAC35932.10
4-Ethylphenylboronsäure 97%
4-Ethylphenylboronsäure
Formel: C₈H₁₁BO₂
Molecular Weight: 149.98 g/mol
Dichte: 1.125
Lagertemperatur: Kühlschrank
MDL: MFCD00859377
CAS-Nummer: 63139-21-9

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR