4-Ethylphenylboronsäure ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (4-Ethylphenyl)metaborsäure, 4-Ethylbenzolboronsäure, Dihydroxy(4-ethylphenyl)boran

B24656.03 B24656.06 B24656.14
B24656.03EA 26.2 EUR
B24656.03 B24656.06 B24656.14
4-Ethylphenylboronsäure ≥97%
4-Ethylphenylboronsäure

Purity: 97% CAS No.: 63139-21-9 Molecular Formula: C8H11BO2 Formula Weight: 149.99 Melting Point: 150-154° Beilstein Reference: 3030638

Formel: C₈H₁₁BO₂
Molecular Weight: 149.98 g/mol
Schmelzpunkt: 150…154 °C
Dichte: 1.125 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00859377
CAS-Nummer: 63139-21-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR