2-Formylphenylboronsäure ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Boronobenzaldehyd, (2-Formylphenyl)metaborsäure, Dihydroxy(2-formylphenyl)boran, 2-Formylbenzolboronsäure

B25434.03 B25434.06
B25434.03EA 46.5 EUR
B25434.03 B25434.06
2-Formylphenylboronsäure ≥97%
2-Formylphenylboronsäure

Purity: 97% CAS No.: 40138-16-7 Molecular Formula: C7H7BO3 Formula Weight: 149.94 Melting Point: ca 115-120° Beilstein Reference: 3030776

Formel: C₇H₇BO₃
Molecular Weight: 149,94 g/mol
Schmelzpunkt: ∼115…120 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00151822
CAS-Nummer: 40138-16-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR