2-Formylphenylboronsäure

Lieferant: FLUOROCHEM

Synonyme: 2-Boronobenzaldehyd, (2-Formylphenyl)metaborsäure, Dihydroxy(2-formylphenyl)boran, 2-Formylbenzolboronsäure

010998-5G 010998-25G
FLUO010998-5GEA 42.9 EUR
FLUO010998-5G FLUO010998-25G
2-Formylphenylboronsäure
2-Formylphenylboronsäure
Formel: C₇H₇BO₃
Molecular Weight: 149.94 g/mol
MDL: MFCD00151822
CAS-Nummer: 40138-16-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR