3-Formylphenylboronsäure

Lieferant: FLUOROCHEM

Synonyme: 3-Boronobenzaldehyd, Dihydroxy(3-formylphenyl)boran, (3-Formylphenyl)metaborsäure, 3-Formylbenzolboronsäure

010798-10G 010798-25G
FLUO010798-10GEA 39.9 EUR
FLUO010798-10G FLUO010798-25G
3-Formylphenylboronsäure
3-Formylphenylboronsäure
Formel: C₇H₇BO₃
Molecular Weight: 149.94 g/mol
MDL: MFCD00161356
CAS-Nummer: 87199-16-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR