4-Formylphenylboronsäure
Lieferant: FLUOROCHEM
|
Achtung
|
Synonyme:
(4-Formylphenyl)metaborsäure, 4-Boronobenzaldehyd, Dihydroxy(4-formylphenyl)boran, 4-Formylbenzolboronsäure
010799-25G
010799-5G
FLUO010799-25GEA
60.5
EUR
FLUO010799-25G
FLUO010799-5G
4-Formylphenylboronsäure
4-Formylphenylboronsäure
Formel:
C₇H₇BO₃ Molecular Weight: 149.94 g/mol |
MDL:
MFCD00151823 CAS-Nummer: 87199-17-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...