1-Brom-3-ethylbenzol

Lieferant: FLUOROCHEM
Achtung

024168-25G 024168-5G
FLUO024168-25GEA 143 EUR
FLUO024168-25G FLUO024168-5G
1-Brom-3-ethylbenzol
1-Brom-3-ethylbenzol
Formel: C₈H₉Br
Molecular Weight: 185,06 g/mol
MDL: MFCD00156128
CAS-Nummer: 2725-82-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR