4-Bromcumol ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Bromoisopropylbenzene, 1-Bromo-4-isopropylbenzene

A13370.06 A13370.09 A13370.18
A13370.06EA 65.4 EUR
A13370.06 A13370.09 A13370.18
4-Bromcumol ≥97%
4-Bromcumol

Purity: 97% CAS No.: 586-61-8 Molecular Formula: C9H11Br Formula Weight: 199.10 Flash Point: 100°(212°F) Refractive Index: 1.5360 Melting Point: -12° Boiling Point: 219° Solubility: Insoluble in water Beilstein Reference: 1858096 UN No.: 3082 MDL No.: MFCD00039159

Formel: C₉H₁₁Br
Molecular Weight: 199,09 g/mol
Siedepunkt: 219 °C
Schmelzpunkt: –12 °C
Dichte: 1,315
Flammpunkt: >100 °C (212 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00039159
CAS-Nummer: 586-61-8
EINECS: 209-577-5
UN: 3082
ADR: 9,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR