1-Brom-4-pentylbenzol ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Brom-pentylbenzol, (4-Bromphenyl)pentan, p-Brom-pentylbenzol

A14457.06 A14457.03
A14457.06EA 88.8 EUR
A14457.06 A14457.03
1-Brom-4-pentylbenzol ≥98%
1-Brom-4-pentylbenzol

Purity: 98% CAS No.: 51554-95-1 Molecular Formula: C11H15Br Formula Weight: 227.15 MDL No.: MFCD00061113

Formel: C₁₁H₁₅Br
Molecular Weight: 227,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00061113
CAS-Nummer: 51554-95-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR