α-Brom-3,4-dichlortoluol

Lieferant: Thermo Scientific
Gefahr

Synonyme: 3,4-Dichlorobenzyl bromide

BTB06044.1 BTB06044.10 BTB06044.50
MAYBBTB06044.1EA 0 EUR
MAYBBTB06044.1 MAYBBTB06044.10 MAYBBTB06044.50
α-Brom-3,4-dichlortoluol
α-Brom-3,4-dichlortoluol
Formel: C₇H₅BrCl₂
Molecular Weight: 239,93 g/mol
Siedepunkt: 84…86 °C (0,3 mmHg)
MDL: MFCD00041363
CAS-Nummer: 18880-04-1
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR