4-Brom-3-chlortoluol

Lieferant: FLUOROCHEM

018263-1G 018263-5G 018263-25G
FLUO018263-1GEA 0 EUR
FLUO018263-1G FLUO018263-5G FLUO018263-25G
4-Brom-3-chlortoluol
4-Brom-3-chlortoluol
Formel: C₇H₆BrCl
Molecular Weight: 205,48 g/mol
MDL: MFCD00045161
CAS-Nummer: 6627-51-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR