3-Brom-4-chlortoluol
Lieferant: FLUOROCHEM
018261-1G
018261-5G
018261-25G
FLUO018261-1GEA
0
EUR
FLUO018261-1G
FLUO018261-5G
FLUO018261-25G
3-Brom-4-chlortoluol
3-Brom-4-chlortoluol
Formel:
C₇H₆BrCl Molecular Weight: 205,48 g/mol |
MDL:
MFCD00079710 CAS-Nummer: 57310-39-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...