4-Brom-2-chlortoluol
Lieferant: FLUOROCHEM
|
Achtung
|
Synonyme:
1-Bromo-3-chloro-4-methylbenzene
018262-5G
018262-25G
018262-100G
FLUO018262-5GEA
0
EUR
FLUO018262-5G
FLUO018262-25G
FLUO018262-100G
4-Brom-2-chlortoluol
4-Brom-2-chlortoluol
Formel:
C₇H₆BrCl Molecular Weight: 205,48 g/mol |
MDL:
MFCD00060651 CAS-Nummer: 89794-02-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...