4-Brom-2-chlortoluol, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 1-Bromo-3-chloro-4-methylbenzene

M-Clarity™ Qualitätsstufe = MQ200

528889-100G 528889-25G
SIAL528889-100GEA 44 EUR
SIAL528889-100G SIAL528889-25G
4-Brom-2-chlortoluol, Sigma-Aldrich®
4-Brom-2-chlortoluol
Formel: C₇H₆BrCl
Molecular Weight: 205,48 g/mol
MDL: MFCD00060651
CAS-Nummer: 89794-02-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR