2-Brom-4-chlortoluol

Lieferant: FLUOROCHEM
Achtung

Synonyme: 1-Brom-3-chlor-6-methylbenzol

018259-10G 018259-25G
FLUO018259-10GEA 0 EUR
FLUO018259-10G FLUO018259-25G
2-Brom-4-chlortoluol
2-Brom-4-chlortoluol
Formel: C₇H₆BrCl
Molecular Weight: 205,48 g/mol
MDL: MFCD00060650
CAS-Nummer: 27139-97-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR