5-Brom-2-chlortoluol
Lieferant: FLUOROCHEM
Synonyme:
1-Brom-4-chlor-3-methylbenzol
018264-10G
018264-25G
FLUO018264-10GEA
33.5
EUR
FLUO018264-10G
FLUO018264-25G
5-Brom-2-chlortoluol
5-Brom-2-chlortoluol
Formel:
C₇H₆BrCl Molecular Weight: 205,48 g/mol |
MDL:
MFCD00018503 CAS-Nummer: 54932-72-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...