ω-Brom-3-chloracetophenon
Lieferant: BIONET RESEARCH
Synonyme:
2-Brom-3'-chloracetophenon, 3-Chlorphenacylbromid, α-Brom-3-chloracetophenon, Brommethyl-m-chlorphenylketon
10X-0707.5
10X-0707.500
10X-0707.10
10X-0707.1
BIOT10X-0707.5EA
0
EUR
BIOT10X-0707.5
BIOT10X-0707.500
BIOT10X-0707.10
BIOT10X-0707.1
ω-Brom-3-chloracetophenon
ω-Brom-3-chloracetophenon
Formel:
C₈H₆BrClO Molecular Weight: 233,49 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00792669 CAS-Nummer: 41011-01-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...