α-Brom-2,4-dichlor-5-fluortoluol
Lieferant: FLUOROCHEM
Synonyme:
2,4-Dichlor-5-fluorbenzylbromid
005731-1G
005731-5G
FLUO005731-1GEA
128
EUR
FLUO005731-1G
FLUO005731-5G
α-Brom-2,4-dichlor-5-fluortoluol
α-Brom-2,4-dichlor-5-fluortoluol
Formel:
C₇H₄BrCl₂F Molecular Weight: 257.92 g/mol |
MDL:
MFCD01631381 CAS-Nummer: 261763-27-3 UN: 3265 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...