α-Brom-2-chlor-6-fluortoluol
Lieferant: FLUOROCHEM
Synonyme:
2-Chlor-6-fluorbenzylbromid
005632-5G
005632-1G
005632-25G
FLUO005632-5GEA
48.8
EUR
FLUO005632-5G
FLUO005632-1G
FLUO005632-25G
α-Brom-2-chlor-6-fluortoluol
α-Brom-2-chlor-6-fluortoluol
Formel:
C₇H₅BrClF Molecular Weight: 223,47 g/mol Siedepunkt: 223 °C (1013 hPa) Schmelzpunkt: 37 °C Dichte: 1,629 g/cm³ (20 °C) |
MDL:
MFCD00040126 CAS-Nummer: 68220-26-8 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...