α-Brom-3-chlor-2,6-difluortoluol

Lieferant: FLUOROCHEM

Synonyme: 3-Chlor-2,6-difluorbenzylbromid

005601-1G 005601-5G
FLUO005601-1GEA 104 EUR
FLUO005601-1G FLUO005601-5G
α-Brom-3-chlor-2,6-difluortoluol
α-Brom-3-chlor-2,6-difluortoluol
Formel: C₇H₄BrClF₂
Molecular Weight: 241,46 g/mol
Siedepunkt: 218 °C (1013 hPa)
Schmelzpunkt: 50 °C
MDL: MFCD01631326
CAS-Nummer: 261762-47-4
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR