α-Brom-3-chlor-4-fluortoluol
Lieferant: FLUOROCHEM
Synonyme:
3-Chlor-4-fluorbenzylbromid
005633-25G
005633-100G
FLUO005633-25GEA
55.4
EUR
FLUO005633-25G
FLUO005633-100G
α-Brom-3-chlor-4-fluortoluol
α-Brom-3-chlor-4-fluortoluol
Formel:
C₇H₅BrClF Molecular Weight: 223.47 g/mol Siedepunkt: 223 Schmelzpunkt: 37 |
MDL:
MFCD01631551 CAS-Nummer: 192702-01-5 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...