α-Brom-5-chlor-2-fluortoluol
Lieferant: FLUOROCHEM
Synonyme:
5-Chlor-2-fluorbenzylbromid
005634-1G
005634-5G
FLUO005634-1GEA
42.9
EUR
FLUO005634-1G
FLUO005634-5G
α-Brom-5-chlor-2-fluortoluol
α-Brom-5-chlor-2-fluortoluol
Formel:
C₇H₅BrClF Molecular Weight: 223.47 g/mol Siedepunkt: 44…45 °C (0.5 mmHg) |
MDL:
MFCD01631437 CAS-Nummer: 71916-91-1 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...