α-Brom-4-chlor-2-fluortoluol
Lieferant: FLUOROCHEM
Synonyme:
4-Chlor-2-fluorbenzylbromid
009420-5G
009420-25G
FLUO009420-5GEA
31.1
EUR
FLUO009420-5G
FLUO009420-25G
α-Brom-4-chlor-2-fluortoluol
α-Brom-4-chlor-2-fluortoluol
Formel:
C₇H₅BrClF Molecular Weight: 223.47 g/mol |
MDL:
MFCD00831105 CAS-Nummer: 71916-82-0 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...