α-Brom-4-chlor-3-fluortoluol

Lieferant: FLUOROCHEM

Synonyme: 4-Chlor-3-fluorbenzylbromid

021808-5G 021808-25G
FLUO021808-5GEA 62.9 EUR
FLUO021808-5G FLUO021808-25G
α-Brom-4-chlor-3-fluortoluol
α-Brom-4-chlor-3-fluortoluol
Formel: C₇H₅BrClF
Molecular Weight: 223.47 g/mol
MDL: MFCD04115859
CAS-Nummer: 206362-80-3
UN: 3261
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR