2'-Bromacetophenon 98+%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
o-Bromacetophenon
179760250
179760050
179761000
ACRO179760250EA
92.8
EUR
ACRO179760250
ACRO179760050
ACRO179761000
2'-Bromacetophenon 98+%
2'-Bromacetophenon
Formel:
C₈H₇BrO Molecular Weight: 199.05 g/mol Siedepunkt: 116…117 °C (12 mmHg) Schmelzpunkt: 0 Dichte: 1.476 g/cm³ Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000067 CAS-Nummer: 2142-69-0 EINECS: 218-398-1 |
Spezifikation Testergebnisse
Appearance | Clear light yellow to orange Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5650 to 1.5690 (20°C, 589 nm) |
GC | ≥98.0 % |
Residue after evaporation | ≤0.01 % |
Iron (Fe) | ≤0.2 ppm |
Lead (Pb) | ≤0.02 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...