ω,3-Dibromacetophenon

Lieferant: MENAI

Synonyme: 2,3'-Dibromacetophenon, 2-Brom-1-(3-bromphenyl)-1-ethanon, 3-Bromphenacylbromid, α,3-Dibromacetophenon

ST2461.10 ST2461.100
MENAST2461.10EA 0 EUR
MENAST2461.10 MENAST2461.100
ω,3-Dibromacetophenon
ω,3-Dibromacetophenon
Formel: C₈H₆Br₂O
Molecular Weight: 277.94 g/mol
Schmelzpunkt: 51
MDL: MFCD00082698
CAS-Nummer: 18523-22-3
UN: 3261
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR