ω,4-Dibromacetophenon, Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
Synonyme:
2-Brom-1-(4-bromphenyl)-1-ethanon, Tau Protein Kinase I Inhibitor, 2,4'-Dibromacetophenon, Glycogen Synthase Kinase-3β Inhibitor VII, GSK-3β Inhibitor VII, 4-Bromphenacylbromid, TPK I Inhibitor, α,4-Dibromacetophenon
M-Clarity™ Qualitätsstufe = MQ200
D38308-100G
D38308-10G
D38308-50G
SIALD38308-100GEA
136
EUR
SIALD38308-100G
SIALD38308-10G
SIALD38308-50G
ω,4-Dibromacetophenon, Sigma-Aldrich®
ω,4-Dibromacetophenon
Formel:
C₈H₆Br₂O Molecular Weight: 277,94 g/mol Siedepunkt: 310 °C (1013 hPa) Schmelzpunkt: 107…108 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00000200 CAS-Nummer: 99-73-0 EINECS: 202-783-6 UN: 3261 ADR: 8,II Merck Index: 13,01412 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...