2-Brom-m-toluylsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

560219-1G 560219-5G
SIAL560219-1GEA 26 EUR
SIAL560219-1G SIAL560219-5G
2-Brom-m-toluylsäure, Sigma-Aldrich®
2-Brom-m-toluylsäure
Formel: C₈H₇BrO₂
Molecular Weight: 215,05 g/mol
MDL: MFCD00079721
CAS-Nummer: 53663-39-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR