6-Brom-o-toluylsäure ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Bromo-6-methylbenzoic acid
L20149.03
L20149.06
L20149.03EA
87.6
EUR
L20149.03
L20149.06
6-Brom-o-toluylsäure ≥98%
6-Brom-o-toluylsäure
Purity: 98%
CAS No.: 90259-31-7
Molecular Formula: C8H7BrO2
Formula Weight: 215.05
Melting Point: 107-111°
Beilstein Reference: 2614094
Formel:
C₈H₇BrO₂ Molecular Weight: 215,05 g/mol Schmelzpunkt: 107…111 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01310788 CAS-Nummer: 90259-31-7 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...