4-Brom-o-toluylsäure, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
4-Bromo-2-methylbenzoic acid
M-Clarity™ Qualitätsstufe = MQ100
665126-100G
665126-25G
665126-5G
SIAL665126-100GEA
524
EUR
SIAL665126-100G
SIAL665126-25G
SIAL665126-5G
4-Brom-o-toluylsäure, Sigma-Aldrich®
4-Brom-o-toluylsäure
Formel:
C₈H₇BrO₂ Molecular Weight: 215,05 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00040905 CAS-Nummer: 68837-59-2 EINECS: 272-437-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...