2,4,6-Trifluorbenzylbromid

Lieferant: FLUOROCHEM
Gefahr

Synonyme: α-Brom-2,4,6-trifluortoluol

007506-1G 007506-5G
FLUO007506-1GEA 42.9 EUR
FLUO007506-1G FLUO007506-5G
2,4,6-Trifluorbenzylbromid
2,4,6-Trifluorbenzylbromid
Formel: C₇H₄BrF₃
Molecular Weight: 225.01 g/mol
MDL: MFCD00061210
CAS-Nummer: 151411-98-2
UN: 3265
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR