3,4,5-Trifluorbenzylbromid
Lieferant: FLUOROCHEM
|
Gefahr
|
Synonyme:
α-Brom-3,4,5-trifluortoluol
007507-1G
007507-5G
FLUO007507-1GEA
39.9
EUR
FLUO007507-1G
FLUO007507-5G
3,4,5-Trifluorbenzylbromid
3,4,5-Trifluorbenzylbromid
Formel:
C₇H₄BrF₃ Molecular Weight: 225.01 g/mol Siedepunkt: 44…46 °C (1 mmHg) Dichte: 1.689 g/cm³ (20 °C) |
MDL:
MFCD00083528 CAS-Nummer: 220141-72-0 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...