3-Brom-α,α,α,2-tetrafluortoluol
Lieferant: FLUOROCHEM
Synonyme:
1-Brom-2-fluor-3-(trifluormethyl)benzol, 3-Brom-2-fluorbenzotrifluorid
CFF577.5
004214-1G
004214-5G
CHMPCFF577.5EA
666
EUR
CHMPCFF577.5
FLUO004214-1G
FLUO004214-5G
3-Brom-α,α,α,2-tetrafluortoluol
3-Brom-α,α,α,2-tetrafluortoluol
Formel:
C₇H₃BrF₄ Molecular Weight: 243 g/mol |
MDL:
MFCD00070812 CAS-Nummer: 144584-67-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...