3-Brom-α,α,α,4-tetrafluortoluol

Lieferant: CHEMPUR
Achtung

Synonyme: 3-Brom-4-fluorbenzotrifluorid, 1-Brom-2-fluor-5-(trifluormethyl)benzol

FL4215.1 004215-5G 004215-25G 004215-100G
CHMPFL4215.1EA 0 EUR
CHMPFL4215.1 FLUO004215-5G FLUO004215-25G FLUO004215-100G
3-Brom-α,α,α,4-tetrafluortoluol
3-Brom-α,α,α,4-tetrafluortoluol
Formel: C₇H₃BrF₄
Molecular Weight: 243 g/mol
MDL: MFCD00040938
CAS-Nummer: 68322-84-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR