2-Brom-α,α,α,3-tetrafluortoluol
Lieferant: FLUOROCHEM
Synonyme:
2-Brom-3-fluorbenzotrifluorid, 1-Brom-2-fluor-6-(trifluormethyl)benzol
004212-1G
004212-5G
FLUO004212-1GEA
36.9
EUR
FLUO004212-1G
FLUO004212-5G
2-Brom-α,α,α,3-tetrafluortoluol
2-Brom-α,α,α,3-tetrafluortoluol
Formel:
C₇H₃BrF₄ Molecular Weight: 243 g/mol |
MDL:
MFCD00061243 CAS-Nummer: 104540-42-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...