2-Brom-α,α,α,6-tetrafluortoluol

Lieferant: FLUOROCHEM

Synonyme: 2-Brom-6-fluorbenzotrifluorid, 1-Brom-3-fluor-2-(trifluormethyl)benzol

005820-1G 005820-2.5G
FLUO005820-1GEA 31.1 EUR
FLUO005820-1G FLUO005820-2.5G
2-Brom-α,α,α,6-tetrafluortoluol
2-Brom-α,α,α,6-tetrafluortoluol
Formel: C₇H₃BrF₄
Molecular Weight: 243 g/mol
MDL: MFCD01631569
CAS-Nummer: 261951-85-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR