4-Brom-α,α,α,2-tetrafluortoluol

Lieferant: CHEMPUR
Achtung

Synonyme: 4-Brom-2-fluorbenzotrifluorid, 1-Brom-3-fluor-4-(trifluormethyl)benzol

FL13780.POR 013780-5G 013780-1G 013780-25G
CHMPFL13780.POREA 0 EUR
CHMPFL13780.POR FLUO013780-5G FLUO013780-1G FLUO013780-25G
4-Brom-α,α,α,2-tetrafluortoluol
4-Brom-α,α,α,2-tetrafluortoluol
Formel: C₇H₃BrF₄
Molecular Weight: 243 g/mol
MDL: MFCD01075254
CAS-Nummer: 142808-15-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR