3-Brom-α,α,α,5-tetrafluortoluol
Lieferant: FLUOROCHEM
004216-5G
004216-25G
FLUO004216-5GEA
0
EUR
FLUO004216-5G
FLUO004216-25G
3-Brom-α,α,α,5-tetrafluortoluol
3-Brom-α,α,α,5-tetrafluortoluol
Formel:
C₇H₃BrF₄ Molecular Weight: 243 g/mol |
MDL:
MFCD00042498 CAS-Nummer: 130723-13-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...