2-Brom-α,α,α,5-tetrafluortoluol

Lieferant: FLUOROCHEM

004213-25G 004213-100G 004213-5G
FLUO004213-25GEA 31.1 EUR
FLUO004213-25G FLUO004213-100G FLUO004213-5G
2-Brom-α,α,α,5-tetrafluortoluol
2-Brom-α,α,α,5-tetrafluortoluol
Formel: C₇H₃BrF₄
Molecular Weight: 243 g/mol
MDL: MFCD00040937
CAS-Nummer: 40161-55-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR