ω-Brom-4-fluoracetophenon

Lieferant: FLUOROCHEM

Synonyme: 2-Brom-1-(4-fluorphenyl)ethan-1-on, Brommethyl-p-fluorphenylketon, 2-Brom-4'-fluoracetophenon, 4-Fluorphenacylbromid, α-Brom-4-fluoracetophenon

018027-5G 018027-25G 018027-100G
FLUO018027-5GEA 36.9 EUR
FLUO018027-5G FLUO018027-25G FLUO018027-100G
ω-Brom-4-fluoracetophenon
ω-Brom-4-fluoracetophenon
Formel: C₈H₆BrFO
Molecular Weight: 217,04 g/mol
Siedepunkt: 82 °C (0,4 mmHg)
Schmelzpunkt: 49 °C
MDL: MFCD00040830
CAS-Nummer: 403-29-2
EINECS: 206-955-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR