4-Brom-3-nitrobenzotrifluorid

Lieferant: FLUOROCHEM

Synonyme: 4-Brom-3-nitrobenzotrifluorid, 1-Brom-2-nitro-4-(trifluormethyl)benzol

002194-100G 002194-25G
FLUO002194-100GEA 171 EUR
FLUO002194-100G FLUO002194-25G
4-Brom-3-nitrobenzotrifluorid
4-Brom-3-nitrobenzotrifluorid
Formel: C₇H₃BrF₃NO₂
Molecular Weight: 270.01 g/mol
Siedepunkt: 102…105 °C (10 mmHg)
Dichte: 1.76 g/cm³ (20 °C)
MDL: MFCD00014685
CAS-Nummer: 349-03-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR