3-Brom-α,α,α-trifluor-5-nitrotoluol

Lieferant: CHEMPUR

FL13787.POR 013787-1G
CHMPFL13787.POREA 0 EUR
CHMPFL13787.POR FLUO013787-1G
3-Brom-α,α,α-trifluor-5-nitrotoluol
3-Brom-α,α,α-trifluor-5-nitrotoluol
Formel: C₇H₃BrF₃NO₂
Molecular Weight: 270,01 g/mol
MDL: MFCD03788719
CAS-Nummer: 630125-49-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR