2-Brom-α,α,α-trifluor-5-nitrotoluol

Lieferant: FLUOROCHEM

Synonyme: 2-Brom-5-nitrobenzotrifluorid, 1-Brom-4-nitro-2-(trifluormethyl)benzol

004256-5G 004256-10G 004256-50G
FLUO004256-5GEA 0 EUR
FLUO004256-5G FLUO004256-10G FLUO004256-50G
2-Brom-α,α,α-trifluor-5-nitrotoluol
2-Brom-α,α,α-trifluor-5-nitrotoluol
Formel: C₇H₃BrF₃NO₂
Molecular Weight: 270,01 g/mol
Siedepunkt: 125…130 °C (12 mmHg)
Schmelzpunkt: 46…48 °C
MDL: MFCD00014707
CAS-Nummer: 367-67-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR