α-Brom-3-(difluormethoxy)toluol

Lieferant: FLUOROCHEM
Gefahr

Synonyme: 3-(Difluormethoxy)benzylbromid, m-(Difluormethoxy)benzylbromid

007862-1G 007862-5G
FLUO007862-1GEA 60.9 EUR
FLUO007862-1G FLUO007862-5G
α-Brom-3-(difluormethoxy)toluol
α-Brom-3-(difluormethoxy)toluol
Formel: C₈H₇BrF₂O
Molecular Weight: 237,04 g/mol
MDL: MFCD00236232
CAS-Nummer: 72768-95-7
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR