5-Brom-2-iodtoluol ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 1-Bromo-4-iodo-3-methylbenzene

B21878.14 B21878.22
B21878.14EA 56.6 EUR
B21878.14 B21878.22
5-Brom-2-iodtoluol ≥98%
5-Brom-2-iodtoluol

Purity: 98+% CAS No.: 116632-39-4 Molecular Formula: C7H6BrI Formula Weight: 296.93 Flash Point: 110°(230°F) Refractive Index: 1.6515 Boiling Point: 262-265° Beilstein Reference: 2325157 MDL No.: MFCD00060664

Formel: C₇H₆BrI
Molecular Weight: 296.93 g/mol
Siedepunkt: 262…265 °C
Dichte: 2.139
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00060664
CAS-Nummer: 116632-39-4
EINECS: 629-570-4

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR